Hygiene mask
Supplier: Hubei Orient
113-8793EA 57.9 EUR
113-8793
Hygiene mask
Masks Hygiene Masks
This disposable face mask is made from 100% non woven PP fabric.
  • Three-ply
  • Integrated nose clip
  • Tie type
  • For industrial and clean room use only; do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
  • This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425
Balení: Supplied in a dispenser box with 50 pieces.