Steam sterilisation indicators
Supplier: Südpack Medical
489-0100EA 217 EUR
489-0100
Steam sterilisation indicators
Sterilisation Indicators
Steam sterilisation. The labels change colour from pink to dark brown after sterilisation.
Balení: Roll of 5000 labels.