Digital Solutions
Applications
Support
About
Order Entry
Disposable particle filtering respirators, FFP3, healthcare, Aura™ 1883+
111-1525
:
3M
Disposable particle filtering respirators, FFP3, healthcare, Aura™ 1883+
111-1525
:
3M
93,10 €
1 * 8 items
-
+
Type:
1883+
Size:
U*
Environmentally Preferable:
Pk:
8
With valve:
+
Class:
FFP3