You Searched For: Manifolds
40  results were found
SearchResultCount:"40"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (8095-06)
Supplier: Avantor
Description: The BAKERBOND Speedisk® expanded extraction station can be used in reservoir, inverted, or remote sample feed modes. It includes a six-port vacuum man...
UOM: 1 * 1 items
Catalog Number: (535-0391)
Supplier: LABCONCO
Description: 8-port manifold, W×D×H: 218×218×399 mm
UOM: 1 * 1 items
Sustainable Materials
Sustainable Packaging
Supplier: Sartorius
Description: Microsart® Manifolds accurately detect and quantify microorganisms in liquid samples. Simply select the proper manifold for your individual microbiolo...
Supplier: Sartorius
Description: The Combisart® system enables you to select the optimal hardware and consumables for your needs in quality assurance. It features a modular design and...
Sale
Supplier: ENTEGRIS INC
Description: Stainless steel/PFA manifold for use with Optimizer® ST DEV and DI photochemical filters.
Supplier: Cytiva (Formerly Pall Lab)
Description: The multi-well plate vacuum manifold is an anodised aluminium manifold that has been designed and optimised for the vacuum filtration of AcroPrepâ„¢, Ac...
New Product
Catalog Number: (SCTV106660)
Supplier: S.C.A.T.
Description: This purging manifold alows you to install up to four SafetyCaps on your purging bottle.
UOM: 1 * 1 items
Supplier: Bohlender
Description: Block made of PTFE with one inlet and four outlets with female thread UNF ¼" 28 G.
Catalog Number: (ALEXADI-987-500)
Supplier: Enzo Life Sciences
Description: The manifold is constructed of a chemical-resistant polymer and is designed to facilitate the filtration of liquid through a multi-well filter plate.
UOM: 1 * 1 items
Catalog Number: (GILI37012000)
Supplier: GILSON LCMS
Description: The ASPEC® Positive pressure manifold uses positive pressure to improve reproducibility and recovery for solid phase extraction (SPE) applications.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: Microtest manifold, stainless steel, 12-position, straight, 96 well, 9 mm centre-to-centre
Catalog Number: (516-1038)
Supplier: Cytiva (Formerly Pall Lab)
Description: This microbiology manifold, with modular design, allows you to optimise testing without sacrificing cleanliness.
UOM: 1 * 1 items
Catalog Number: (ENTEDZUM044C1)
Supplier: ENTEGRIS INC
Description: Stainless steel manifold for use with Impact® Plus disposable filters.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: HyperSepâ„¢, Positive pressure manifold, with 13 mm collection rack
Supplier: Thermo Fisher Scientific
Description: HyperSepâ„¢ vacuum manifolds are designed for use with HyperSepâ„¢ SPE columns and HyperSepâ„¢ 96-well plates.
Supplier: Thermo Fisher Scientific
Description: Super Cleanâ„¢, Cartridge filter base plate, single base. For carrier gas only, for triple filter
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 40
no targeter for Bottom