Portable water sampler, TP 5 P
Supplier: Maxx
701-0167EA 2750 EUR
MAXA2051000
Portable water sampler, TP 5 P
Robust, battery operated sampler that can be combined with any composite sample container as well as with the optional passive or active insulating boxes. The sample collection is automatic with a diaphragm pump vacuum system using simple programmable microprocessor control with an adjustable 20 to 350 ml sample volume system.
  • Very tough plastic housing
  • Highly mobile device and independent of mains power with an integral 12 V battery
  • Programming can be time-, flow- or event-related
  • Manual sample extraction without interruption of program
  • 2000 sample extractions per battery charge
Informace pro objednávání: For details of the range of accessories, please visit www.vwr.com or contact your local Avantor sales office.
Delivery information: The device (without battery charger) includes 5 m suction hose with threaded connection and sinker weight.