Anti-AGR3 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-6508R-CY3EA 858 EUR
BOSSBS-6508R-CY3
Anti-AGR3 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
BCMP11

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGR3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR