Anti-ABCA2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BOSSBS-6332R-CY5.5EA 858 EUR
BOSSBS-6332R-CY5.5
Anti-ABCA2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. This protein is highly expressed in brain tissue and may play a role in macrophage lipid metabolism and neural development. Two transcript variants encoding different isoforms have been found for this gene (from EntrezGene).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR