Anti-SLC2A10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6325R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 2 Member A10
- Antigen symbol:SLC2A10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:81031
- Antigen synonyms:GLUT10|ATS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:299-316/541
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Defects in SLC2A10 are the cause of arterial tortuosity syndrome (ATS) [MIM:208050]. ATS is an autosomal recessive disorder characterized by tortuosity and elongation of major arteries, often resulting in death at young age. Other typical features include aneurysms of large arteries and stenosis of the pulmonary artery, in association with facial features and several connective tissue manifestations such as soft skin and joint laxity. Histopathological findings include fragmentation of elastic fibers in the tunica media of large arteries.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC2A10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat