Anti-RASGRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BOSSBS-3382R-A647EA 885 EUR
BOSSBS-3382R-A647
Anti-RASGRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. The studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq, Mar 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASGRF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR