Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Leak Detection Systems
https://si.vwr.com/stibo/smallweb/std.lang.all/52/20/21655220.jpg
SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"12"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (PERCN9306074)
Supplier:  PERKIN ELMER GC-INSTRUMENTS
Description:   The MiniTemp MT4 is capable of displaying thermal measurement readings using easy point and shot infrared technology in a pocket size configuration.
UOM:  1 * 1 items
Catalog Number: (PERCN9306089)
Supplier:  PERKIN ELMER GC-INSTRUMENTS
Description:   Designed for use with gas chromatography (GC) systems, it detects minute leaks of any gas with a thermal conductivity different from air.
UOM:  1 * 1 items
Catalog Number: (HYPE66002-001)
Supplier:  Thermo Scientific
Description:   This handheld instrument can detect minute leaks of laboratory gases.
UOM:  1 * 1 items
Catalog Number: (TEST0563.3165)
Supplier:  Testo
Description:   Constantly checks the level of ammonia for common refrigerants: When leaks are detected, the display changes from green to red.
UOM:  1 * 1 items
Catalog Number: (TEST0560.5350)
Supplier:  Testo
Description:   This carbon dioxide meter comes with a permanent sensor to specifically measure and optimise the ambient air.
UOM:  1 * 1 items
Catalog Number: (TEST0632.0316)
Supplier:  Testo
Description:   Audible, visual 360° flashing LED. Max. 200 ppm LED: Yellow - Max. 10000 ppm LED: Red, More than five hours, Alkali manganese IEC 6LR61, −20…+50
UOM:  1 * 1 items
Catalog Number: (TEST0632.3162)
Supplier:  Testo
Description:   Gas detection, Optical and audible alarm, 6 hours (rechargeable), NiMH, 20 - 80, -5...+50
UOM:  1 * 1 items
Catalog Number: (151-0208)
Supplier:  DRAEGER
Description:   A small heating element in the device head heats an alcohol mixture with a high molecular weight and density similar to air.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 12
  1  2  Next