You Searched For: Cooling Racks
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Corning
Description: These thermoconductive modules provide reproducible and standardised temperature control to biological samples while on the bench.
Catalog Number: (BELAF189050050)
Supplier: Bel-Art Products, a Part of SP
Description: PP
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: Molded of grey high-impact polystyrene.
Catalog Number: (KOEHA-1070)
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Description: PCR vials may be kept at a temperature of 0 Â°C for at least 30 min if the cooler is cooled down to –20 Â°C for approximately 12 h.
UOM: 1 * 1 items
Catalog Number: (DELTM-572)
Supplier: DELTALAB
Description: PP, with clear lid.
UOM: 1 * 2 items
Supplier: DELTALAB
Description: Styrofoam, white
Supplier: DELTALAB
Description: PC.
Supplier: GLW
Description: These cooling racks are made of polypropylene for test tubes that require continued cooling.
Supplier: SARSTEDT
Description: Isofreeze® racks are for use as a pipetting and storage station for temperature sensitive applications like enzyme analyses, PCR or cell based assays....
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom