You Searched For: Multi-Parameter Electrodes
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (HACHLZW5059970002)
Supplier: Hach
Description: sensION+ 5059 is a combination multi-parameter electrode for determination of pH, conductivity and temperature. It has a plastic body, non-refillable ...
UOM: 1 * 1 items
Catalog Number: (HACHLZW5045970002)
Supplier: Hach
Description: sensION+ 5045 is a combination multi-parameter electrode for determination of pH, ORP and temperature. Dedicated for use with Hach sensION+ MM110 port...
UOM: 1 * 1 items
Catalog Number: (HACHLZW5048970002)
Supplier: Hach
Description: Multisensor for pH, ORP, conductivity and temperature. For use with sensION+ MM150 multi-parameter meter.
UOM: 1 * 1 items
Supplier: Endress+Hauser
Description: The CYM17 Memosens-analog converter enables easy use of digital Memosens sensors in your fermentation applications in the laboratory without changing ...
New Product
Catalog Number: (665-0088)
Supplier: YSI (ENVIRONMENTAL PRODUCTS
Description: Electrode, Multi-Parameter
UOM: 1 * 1 items
Catalog Number: (EUTEECCOMBI03M)
Supplier: EUTECH
Description: Combined pH electrode and 2-pin stainless steel conductivity electrode with 15 cm ABS guard. For use with CyberScan PC 10.
UOM: 1 * 1 items
Catalog Number: (EUTE01X001530)
Supplier: EUTECH
Description: Electrode, Multi-Parameter
UOM: 1 * 1 items
Supplier: Cole-Parmer
Description: Electrode, Multi-Parameter
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom