Magnetic hotplate stirrer, RCT classic IKAMAG® safety control with VT-5 contact thermometer
Supplier: IKA
IKAMAG®
442-0599EA 0 EUR
442-0599 442-0600 442-0598
Magnetic hotplate stirrer, RCT classic IKAMAG® safety control with VT-5 contact thermometer
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrer with aluminium alloy plate, suitable for unsupervised operation. It features integrated temperature control, digital error code display and incorporates excellent safety features, such as a 'hot' top indicator to prevent burns and digital display of the set safety temperature limit. With its improved heating control this magnetic hotplate stirrer is ideal for use with contact thermometers, for precise temperature control.
  • Adjustable safety circuit for heating plate temperature (50 to 360 °C)
  • Digital display shows exact temperature and speed setting, even when switched off
  • Bushing according to DIN 12878 for connecting contact thermometer e.g. VT-5 (supplied free of charge)
  • High magnetic adhesion prevents decoupling
  • Can be used at 0 to 60 °C (80% relative humidity), in an incubator or cold room
Certifikace: IP protection class according to DIN EN 60529: IP 42 (RCT classic) and IP 54 (VT-5)
Delivery information: Supplied with VWR® VT-5 contact thermometer and protective cover. A wide range of accessories are available, please order these separately.