Erlenmeyer flasks, with standard ground joint
201-1340EA 19 EUR
VWRI201-1340
Erlenmeyer flasks, with standard ground joint
Borosilicate glass, grade 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The good value alternative for a wide range of laboratory applications. Volume graduations and labelling area.
Certifikace: DIN 12387