You Searched For: Electrodes
333  results were found
SearchResultCount:"333"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These pH electrodes are available with refillable electrolyte or non refillable.
Supplier: VWR Collection
Description: pH electrodes with 1 m fixed cable or S7 connector.
Supplier: VWR Collection
Description: Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.
Supplier: VWR Collection
Description: With 1 m fixed cable.
Supplier: VWR Collection
Description: Electrode, Dissolved Oxygen, VWR®, 10 m cable
Supplier: VWR Collection
Description: Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters...
Supplier: VWR Collection
Description: VWR®, RD 223 (redox), Glass, Ø×L: 12×120 mm, BNC, 1 m cable
Supplier: VWR Collection
Description: Combined pH electrodes with 1 m fixed cable.
Supplier: VWR Collection
Description: Electrode, Conductivity, VWR®
Supplier: VWR Collection
Description: pH electrodes with BNC connector and cable.
Supplier: VWR Collection
Description: These IDP (Intelligent Digital Probe) pH/temperature electrodes can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
Catalog Number: (664-0042)
Supplier: VWR Collection
Description: Electrode, Dissolved Oxygen, VWR® PHenomenal®, pHenomenal OXY11-3, galvanic, 3 m cable, 8-pin
UOM: 1 * 1 items
Catalog Number: (664-0182)
Supplier: VWR Collection
Description: This IDP (Intelligent Digital Probe) dissolved oxygen/temperature sensor can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter mete...
UOM: 1 * 1 items
Catalog Number: (662-1165)
Supplier: VWR Collection
Description: Redox electrode with BNC connector and cable.
UOM: 1 * 1 items
Catalog Number: (663-0356)
Supplier: VWR Collection
Description: This IDP (Intelligent Digital Probe) conductivity/temperature cell can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
UOM: 1 * 1 items
Catalog Number: (663-0147)
Supplier: VWR Collection
Description: Electrode, Conductivity, VWR® PHenomenal®, pHenomenal® CO11, 10 ÂµS/cm - 20 mS/cm, 0...+80 °C, Epoxy, Ø×L: 15,3×120 mm, 0,84 cm-1, 8-pin, 1,5 m cable
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 333
no targeter for Bottom