Rotational viscometers, ViscoPad Nova
Supplier: Fungilab
ViscoPad Nova
616-0115EA 5890 EUR
616-0115 616-0120 616-0119 616-0118 616-0117 616-0116
Rotational viscometers, ViscoPad Nova
Viscometers Rotational Viscometers
The ViscoPad Nova is controlled using an intuitive mobile application on an eight-inch tablet.
  • Data includes speed, viscosity reading, shear rate, shear stress and density
  • Programmable Step - Multistep - Ramp, Time To Torque (TTT) and Time To Stop (TTS) options
  • Data in a comprehensive database for testing purposes
  • Customisation options include speeds, Quality Control Limits (QCL) and out of measurement range alarms
  • Systems with or without PPR (Push, Plug and Release) coupling system
(L) Low to medium viscosity - (R) Medium to high viscosity - (H) High to very high viscosity
Delivery information: Complete with standard spindles (4 for L model and 6 for H and R models), spindle protector, carrying case, USB cable and datalogger software.