Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (5)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Blades and Handles
SearchPresentationType-VERTICAL
SearchResultCount:"12"
  List View Grid View Easy View (new)
Sort by:
Supplier:  LEICA HISTOLOGY
Description:   With these blades it is easy to section a wide range of tissues. Edge encourages immediate ribboning. Premium edge geometry with 35° angle, so no adju...
Supplier:  LEICA MICROSYSTEMS
Description:   These ultra fine, durable disposable blades are made of stainless steel and can be used with the Leica microtome and cryostat series as well as for co...
Supplier:  LEICA MICROSYSTEMS
Description:   Special knife, 50°, Steel, Length: 16 cm
Catalog Number: (PERS72-0114-0000)
Supplier:  PERSONNA MEDICAL
Description:   Personna Plus® microtome blades with MicroCoat® coating.
UOM:  1 * 50 items
Supplier:  FEATHER
Description:   Made of stainless steel. FEATHER® disposable microtome blades are used for paraffin and cryostat sectioning for the whole range of individual applicat...
Supplier:  FEATHER
Description:   These trimming blades in conjunction with the handle F-1326P allow fast and easy blade change thus providing users with increased safety and comfort i...
Supplier:  FEATHER
Description:   The carbon steel trimming blades are special blades for cutting tissue samples without causing deformations. The trimming blade plastic handles are ma...
Catalog Number: (FEAT207000160)
Supplier:  FEATHER
Description:   One-touch microtome blade holders for use with disposable microtome blades for paraffin and cryostat sectioning in a wide range of individual applicat...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 12
  1  2  Next