Anti-TH Rabbit Polyclonal Antibody
NOVUNB300-109
- Antibody type:Primary
- Antigen name:Tyrosine Hydroxylase
- Antigen symbol:TH
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Antigen synonyms:TH-4|tyrosine hydroxylase|EC 1.14.16.2|TYH|Tyrosine 3-hydroxylase|Tyrosine 3-monooxygenase|TY3H|TH isoform 3|TH isoform a
- Size:100 μl
- Pk:0,1 mL
Rabbit Polyclonal Tyrosine Hydroxylase Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Human, Mouse, Rat, Mammal.
There are 2 scientific publications using this antibody.
Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human