Upright microscope, DM300
Supplier: Leica
630-3240EA 1340 EUR
630-3240 630-3393 630-3239 630-3238 630-3105
Upright microscope, DM300
Microscopes
For slightly more advanced students in biology, or 2 to 4 year college life science classrooms, the Leica DM300 compound microscope is compact, maintenance-free and easy to use to provide trouble-free operation each day.
  • Maintenance-free, sealed brass shaft provides accurate focusing control
  • Leica Elzie™ LED illumination provides a cool white light at 80% lower energy consumption
  • LED illumination also eliminates the need to change any bulbs
  • Pre-centred, pre-focused professional condenser system for maximising illumination
  • Clearly labelled adjustable iris diaphragm for fine tuning the image resolution
  • Built-in mechanical stage with low position X/Y controls for accurate specimen movement at high magnifications
  • Compact size with small footprint for easy storage and carrying
Equipped with a mechanical stage to accommodate two specimen slides and labelled Abbe condenser, the DM300 can be configured with rotatable monocular, binocular, or trinocular tubes for shared viewing and easy storage.
The trinocular tube features a 50%/50% camera/eyetube split.
Informace pro objednávání: Power cord is not included (please order separately). 2 year warranty.
Delivery information: Microscope DM300, set 1: Brightfield 3 objective Achromat monocular standard outfit, right hand stage stand with 4-position nosepiece.
Microscope DM300, set 2: Brightfield 4 objective Achromat Monocular Standard Outfit includes DM 300 right hand stage stand with 4-position nosepiece, universal power supply, mechanical stage with slide holder, pre-centred and pre-focused Abbe condenser.
Microscope DM300, set 3: 30° viewing angle, with stand, mechanical stage, Abbe condenser, 4 achromatic objectives (4×, 10×, 40×, 100×), EZ tube with 10×/18 eyepiece and pointer in the right eyepiece, EZLITE LED illumination and dust cover.
Microscope DM300, set 4 : Brightfield 4 objective plan binocular standard outfit includes DM 300 right hand stage stand with 4-position nosepiece, universal power supply, mechanical stage with slide holder, pre-centred and pre-focused Abbe condenser.
Microscope DM300, set 5: 30° viewing angle, with stand, mechanical stage, Abbe condenser, 4 achromatic objectives (4×, 10×, 40×, 100×), EZ tube with 10×/18 eyepiece and pointer in the right eyepiece, EZLITE LED illumination and dust cover.