Full face mask, Sari
Supplier: Scott Safety
Sari
111-2143EA 179.95 EUR
111-2143
Full face mask, Sari
Respirators Reusable Respirators Full Face Respirators
The Sari full face mask is renowned for its high standards of safety, reliability and comfort. Sari is approved for use with Pro2000 filters, with Autoflow and Proflow PAPR powered units and with a constant flow compressed airline.
  • Wide sealing edge for comfort and excellent fit
  • Comfortable to wear even for long periods
  • Adjustable five-point rubber head harness
  • Large distortion-free visor
  • Speech diaphragm affords clear communication
Certifikace: EN 136 Class 3.