You Searched For: Cell Freezing Media
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: SeraFree cryopreservation media is ready to use freezing media for cryopreservation of adherent or suspension cultured cells. The animal-free media co...
Supplier: Charles River Laboratories Cell Solutions
Description: CryoStor® cryopreservation freeze media products have been designed to mitigate temperature-induced molecular cell stress responses during freezing an...
Catalog Number: (HYCLSR30002.02)
Supplier: HyClone products (Cytiva)
Description: HyCloneâ„¢ HyCryo-STEM is a serum-free cryopreservation media designed specifically to maintain differentiation potential and minimise spontaneous diffe...
UOM: 1 * 100 mL
Catalog Number: (SR30001.02)
Supplier: HyClone products (Cytiva)
Description: Animal origin-free HyCloneâ„¢ HyCryo cryopreservation media is designed for general use.
UOM: 1 * 100 mL
Catalog Number: (ICNA0920300M2)
Supplier: MP Biomedicals
Description: Cellvation is a cryopreservation medium designed for optimal cell recovery and convenience.
UOM: 1 * 60 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom