You Searched For: Cell Separation Media
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (D1556-250ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Density gradient medium is suitable for cell and subcellular organelle isolation, virus purification, and isolation of lipoproteins and macromolecules...
UOM: 1 * 250 mL
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particl...
Supplier: VWR Chemicals
Description: Ficoll® 400 is a neutral, hydrophilic polymer of sucrose, which readily dissolves in aqueous solutions.
Supplier: Biowest
Description: Lymphosep is a standardised, high quality medium designed for simple and rapid isolation of PBMCs from treated whole blood. Lymphosep, like Ficollâ„¢, h...
Catalog Number: (734-1431)
Supplier: Corning
Description: Intestinal Epithelium Differentiation Medium (formerly Entero-STIM) promotes the rapid differentiation of intestinal epithelium in vitro.
UOM: 1 * 500 mL
Supplier: Corning
Description: Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
Supplier: Thermo Scientific
Description: Cell separation media, Ficoll® 400
Catalog Number: (734-2765)
Supplier: Corning
Description: Accutase® is an effective cell detachment solution for the routine detachment of cells from standard tissue culture plasticware as well as enhanced su...
UOM: 1 * 100 mL
Catalog Number: (734-1433)
Supplier: Corning
Description: Corning® BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
UOM: 1 * 1 KIT
Catalog Number: (ICNA0216000380)
Supplier: MP Biomedicals
Description: A copolymer of sucrose and epichlorohydrin. Component used to make density gradients for lymphocyte separation.
UOM: 1 * 100 g
Supplier: Cytiva
Description: Ficoll® PM70 is a high molecular weight sucrose polymer formed by copolymerisation of sucrose with epichlorohydrin. The molecules are highly branched ...
Supplier: Cytiva
Description: Ficoll-Paqueâ„¢ PREMIUM products are a range of sterile, ready to use density gradient media for the preparation of mononuclear cells.
Supplier: Cytiva
Description: Percoll® PLUS is for preparation of cells, subcellular particles and larger viruses.
Supplier: MP Biomedicals
Description: Ficoll® is a synthetic, hydrophilic polymer made by the copolymerisation of sucrose and epichlorhydrin. It is a highly branched polymer. Solutions at ...
Supplier: MP Biomedicals
Description: Polysucrose is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relat...
Supplier: Cytiva
Description: Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom