Inhibitors
Catalog Number:
(E633-2KU)
Supplier:
VWR Chemicals
Description:
Human placental source, 40 KU/ml. RNase inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro
UOM:
1 * 2 KU
Supplier:
VWR Chemicals
Description:
A serine protease inhibitor specific for trypsin and trypsin-like proteases.
Supplier:
VWR Chemicals
Description:
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin, papain, kallikrein, thrombin, and cat...
Catalog Number:
(M250-1ML)
Supplier:
VWR Chemicals
Description:
Contains protease inhibitors for mammalian cell use: AEBSF, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin
UOM:
1 * 1 ml
Catalog Number:
(1B1410-2.5KU)
Supplier:
VWR Chemicals
Description:
Recombinant RNase inhibitor purified from E. coli expressing the gene from Sus domesticus.
UOM:
1 * 1 KIT
Catalog Number:
(J581-5MG)
Supplier:
VWR Chemicals
Description:
A reversible cysteine and serine protease inhibitor of trypsin, papain and cathepsin A and B. Slightly inhibits plasmin.
UOM:
1 * 5 mg
Catalog Number:
(M307-1ML)
Supplier:
VWR Chemicals
Description:
Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml di...
UOM:
1 * 1 ml
Catalog Number:
(M183-5MG)
Supplier:
VWR Chemicals
Description:
An aspartic protease inhibitor of pepsin, renin, and cathepsin D. Can also block HIV protease. Not soluble in water.
UOM:
1 * 5 mg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
|
|||||||||