Rotary evaporators, RV 10 control auto
Supplier: IKA
531-0883EA 0 EUR
531-0883 531-0987 531-0886 531-0887 531-0876
Rotary evaporators, RV 10 control auto
Evaporators Rotary Evaporators
Enhanced RV 10 control rotary evaporator with automatic detection of boiling points and direct connection to speed controlled vacuum pump.
  • Increased solvent recovery performance up to 100%
  • Motorised lift with safety stop function
  • Dry run control; safety temperature circuits
  • Users can program, save and run up to 10 individual distilling procedures for greater flexibility
  • Lock function of heating bath settings
  • High performance cooling condenser
  • Reduced noise level and wearing of pump components
The RV 10 control auto offers all features that are necessary for reduced process time and increased overall performance. The evaporator also manages the cooling water flow as it's needed. The gentle maintenance of the vacuum level prevents over- or undershooting of the target vacuum level. It allows for faster, more efficient and safer distilling processes.
Delivery information: Supplied with water/oil heating bath HB 10, speed controlled pump and RV 10.1 Set of glassware (vertical).
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Delivered only with bath, pump and Woulf bottle. This package enables the customer to select the glassware set up for their specific application.