VWR® Clinical Services, Cytotoxic Spill Kits
Supplier: Avantor
121-0150EA 0 EUR
121-0151 121-0150 121-0149 121-0148 121-0147
VWR® Clinical Services, Cytotoxic Spill Kits
Spill Control Materials
Protective clothing, tools, absorbents and wipes arranged in four multifunctional kits, for rapid and safe cleaning of cytotoxic drug spillages. For use in hospitals, clinics, surgeries, pharmacies and laboratories.
Cytotoxic spill kits:
Cytotoxic Spill Kit PPE, Cytotoxic Spill Kit Liquid Clean Up, Cytotoxic Spill Kit Powder Clean Up, Cytotoxic Spill Kit Waste Disposal, and sterile wipes for safe containment and removal of cytotoxic spillages.
Cytotoxic spill kit PPE:
Cleanroom gloves, polychloroprene, BioClean P-Zero™, sterile, size 8, natural (112-3877); chemo protective apron, BioClean-C™, CleanTough™ fabric, size: L, blue (113-8149); sleeve protector, Tyvek®, PS32LA, 500 mm, white (113-0494); cleanroom boot covers, BioClean-D™, CleanTough™, 400 mm, white (113-0814); cleanroom face masks, Maximum with earloop, blue (113-6220); cleanroom goggles, BioClean Clearview™, sterile, single-use, clear PC (111-0186).
Cytotoxic spill kit liquid clean up:
Absorbent, Chemizorb® powder for spillages 500 g (1.02051.0500); autoclavable scraper, PP, L×W: 250x110 mm, green (233-0194); scoop, L×W: 310x138 mm, white (233-0195); chemically inert tweezers, fine square tips, PTFFE, white (232-0211).
Cytotoxic spill kit powder clean up:
Water for injection, Mini Plasco, 20 ml (331-0299); absorbent pad, PIG® HAZ-MAT, heavy weight, 510×380 mm (121-0527).
Cytotoxic spill kit waste disposal:
Disposal bag for hazardous waste, PE, black warning 'Hazardous Material Handle With Care' on bright yellow, small, L×W: 762×457 mm (121-0503); floor sign: 'Caution Hazardous Spill', PVC, L×W: 600×450 mm (116-0112); fixing clamp for floor-standing sign, PVC (116-0116).
Informace pro objednávání: Available as four single kits, and as an 'all-in-one' version (Cytotoxic Spill Kit).