You Searched For: Wrenches
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: Avantor® Hichrom, Combination spanner ³/₈", Stainless steel, Use with head style: ³/₈"
Catalog Number: (18100010.)
Supplier: VWR Chemicals
Description: VWR®, Key for metal drums with 2" and 3/4" openings
UOM: 1 * 1 items
Catalog Number: (HI-199)
Supplier: Avantor
Description: This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
UOM: 1 * 1 items
Catalog Number: (18100150.)
Supplier: VWR Chemicals
Description: VWR®, Key for IBC container, 1000 l capacity, with opening NW 150
UOM: 1 * 1 items
Supplier: VWR Chemicals
Description: A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Catalog Number: (241-0053)
Supplier: VWR Collection
Description: Toughened steel. This six-sided key is designed for use with Labomeca® bosses, joints and feet.
UOM: 1 * 5 items
Supplier: DWK Life Sciences
Description: Applying the correct closure torque to DURAN® caps is critical to achieving an effective seal. Therefore a range of eight specifically designed spline...
Waste Reducing
Supplier: Bohlender
Description: For opening and closing of screw caps.
Catalog Number: (BURK5690-0180)
Supplier: BÃœRKLE
Description: Strap wrench, 180 mm Ø
UOM: 1 * 1 items
Supplier: Bohlender
Description: These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
Catalog Number: (568-0686)
Supplier: SGE a Trajan company
Description: Spanner sizes and uses: 3/16" for use with mini unions, 1/4" for use with 1/16" nuts, 5/16" for use with micro control valves.
UOM: 1 * 1 items
Catalog Number: (BOHLD701-24)
Supplier: Bohlender
Description: These PP ring wrenches can be used for opening and closing multiple distributors for barrels.
UOM: 1 * 1 items
Supplier: Kautex
Description: Wrenches made from HDPE for opening and closing jerricans.
Supplier: IKA
Description: Single head wrench, open, SW8
Catalog Number: (NALG2195-2438)
Supplier: Thermo Fisher Scientific
Description: Durable epoxy resin construction, specifically designed for Nalgene ETFE closures.
UOM: 1 * 1 items
Catalog Number: (119-0238)
Supplier: JUSTRITE MANUFACTURING CO
Description: Drum Wrench opens drum bung caps.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom