You Searched For: Tin Standards
27  results were found
SearchResultCount:"27"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Tin Standard, 1000 mg/L, VWR® AVS TITRINORM, Tin, Matrix: 20% HCl, Application: AAS standards
Catalog Number: (456242U)
Supplier: VWR Chemicals
Description: Sn in HCl.
UOM: 1 * 100 mL
Supplier: Avantor
Description: Tin Standard, 1000 mg/L, J.T.Baker® BAKER INSTRA-ANALYZED®, J.T. Baker®, Tin, Matrix: 2% HNO₃, Application: ICP standards
Catalog Number: (456232S)
Supplier: VWR Chemicals
Description: Sn in HNO₃ tr. HF.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Tin Standard, 1000 mg/L, VWR® ARISTAR®, Tin, Matrix: 2% HNO₃/0,5% HF, Application: ICP-MS standards
Catalog Number: (456252W)
Supplier: VWR Chemicals
Description: Sn in HCl.
UOM: 1 * 100 mL
Supplier: Thermo Scientific
Description: Tin Standard, 10000 mg/L, Specpure®, Matrix: 20% HCl, Application: ICP standards
Supplier: Thermo Scientific
Description: Tin Standard, 1000 mg/L, Specpure®, Matrix: 20% HCl, Application: AAS standards
Catalog Number: (14206.18)
Supplier: Thermo Scientific
Description: Tin Standard, Standard for wear metal analysis, 1000 µg/g, Specpure®, Matrix: oil, Application: Oil based standards
UOM: 1 * 50 g
Catalog Number: (42450.18)
Supplier: Thermo Scientific
Description: Tin Standard, Standard for wear metal analysis, 5000 µg/g, Specpure®, Matrix: Mineral oil, Application: Oil based standards
UOM: 1 * 50 g
Supplier: Thermo Scientific
Description: Tin Standard, 1000 mg/L, Specpure®, Matrix: 20% HCl, Application: ICP standards
Supplier: ACSD
Description: Tin Standard, 10000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: ACSD
Description: Tin Standard, 10000 mg/L, Matrix: 20% HCl, Application: ICP standards
Supplier: ACSD
Description: Tin Standard, 1000 mg/L, Matrix: 20% HCl, Application: ICP standards
Supplier: ACSD
Description: Tin Standard, 1000 mg/L, Matrix: 1% HNO₃/1% HF, Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 27
no targeter for Bottom