You Searched For: Zinc Standards
25  results were found
SearchResultCount:"25"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Zn in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Zinc Standard, 10000 mg/L, VWR®, Zinc, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Catalog Number: (5791.0100)
Supplier: Avantor
Description: Single element plasma standards are used for accuracy and easy of calibration, along with precise detection of trace impurities.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Zn in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Zinc Standard, 1000 mg/L, VWR® AVS TITRINORM, Zinc, Matrix: 2% HNO₃, Application: AAS standards
Supplier: Thermo Scientific
Description: Zinc Standard, Standard for wear metal analysis, 1000 µg/g, Specpure®, Matrix: oil, Application: Oil based standards
Supplier: Thermo Scientific
Description: Zinc Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: AAS standards
Supplier: Thermo Scientific
Description: Zinc Standard, 10000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Catalog Number: (42453.18)
Supplier: Thermo Scientific
Description: Zinc Standard, Standard for wear metal analysis, 5000 µg/g, Specpure®, Matrix: Mineral oil, Application: Oil based standards
UOM: 1 * 50 g
Supplier: Hach
Description: Standard solution for accuracy checks in zinc determinations.
Catalog Number: (PERCN9303758)
Supplier: PerkinElmer
Description: Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
UOM: 1 * 125 mL
Supplier: Thermo Scientific
Description: Zinc Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: ACSD
Description: Zinc Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Supplier: ACSD
Description: Zinc Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Catalog Number: (45289.AD)
Supplier: Thermo Scientific
Description: Zinc Standard, 10 mg/L, Specpure®, Matrix: 2% HNO₃, Application: ICP standards, Isotopic standards
UOM: 1 * 50 mL
Supplier: ACSD
Description: Zinc Standard, 10000 mg/L, Matrix: 5% HCl, Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 25
no targeter for Bottom