You Searched For: Tantalum Standards
16  results were found
SearchResultCount:"16"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Tantalum Standard, 1000 mg/L, VWR® AVS TITRINORM, Matrix: 5% HNO₃, Application: AAS standards
Supplier: VWR Chemicals
Description: Tantalum Standard, 10000 mg/L, VWR® ARISTAR®, Tantalum, Matrix: H₂O/tr HF, Application: ICP standards
Supplier: VWR Chemicals
Description: Tantalum Standard, 1000 mg/L, VWR® ARISTAR®, Tantalum, Matrix: H₂O/tr HF, Application: ICP-MS standards
Catalog Number: (5748.0100)
Supplier: Avantor
Description: Tantalum Standard, 10000 mg/L, J.T.Baker®, J.T. Baker®, Matrix: H₂O/tr HF, Application: ICP standards
UOM: 1 * 100 mL
Supplier: Thermo Scientific
Description: MDL No.: MFCD00011252
Molecular Formula: TaClâ‚… in 2% HF
UN No.: 2922
ADR: 8,II
GHS Symbol(s): GHS06-GHS05
GHS Signal Word: Da...
Supplier: Thermo Scientific
Description: Tantalum Standard, 10000 mg/L, Specpure®, Matrix: 2% HF, Application: ICP standards
Supplier: Thermo Scientific
Description: Tantalum Standard, 1000 mg/L, Specpure®, Matrix: 2% HF, Application: ICP standards
Catalog Number: (PERCN9303803)
Supplier: PerkinElmer
Description: Tantalum Standard, 1000 mg/L, Matrix: Hâ‚‚O/0,8% HF, Application: AAS standards, ICP standards
UOM: 1 * 1 items
Supplier: ACSD
Description: Tantalum Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Supplier: ACSD
Description: Tantalum Standard, 1000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Catalog Number: (ACSDM357.2N05FP.L1)
Supplier: ACSD
Description: Tantalum Standard, 100 mg/L, Matrix: 2% HNO₃/0,5% HF, Application: ICP-MS standards
UOM: 1 * 100 mL
Supplier: ACCUSTANDARD
Description: Tantalum Standard, 10000 mg/L, Matrix: Hâ‚‚O/tr HF, Application: ICP standards
Supplier: ACCUSTANDARD
Description: Tantalum Standard, 1000 mg/L, Matrix: Hâ‚‚O/tr HF, Application: ICP standards
Supplier: LGC Standards PROMOCHEM
Description: These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectro...
Supplier: ACCUSTANDARD
Description: Tantalum Standard, 1000 mg/L, Matrix: Hâ‚‚O/tr HF, Application: AAS standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 16
no targeter for Bottom