You Searched For: Strontium Standards
22  results were found
SearchResultCount:"22"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Strontium Standard, 10 mg/L, VWR® ARISTAR®, Strontium, Matrix: 2% HNO₃, Application: ICP-MS standards
Supplier: Avantor
Description: Strontium Standard, 10000 mg/L, J.T.Baker® BAKER INSTRA-ANALYZED®, J.T. Baker®, Strontium, Matrix: 2% HNO₃, Application: ICP standards
Supplier: VWR Chemicals
Description: Sr(NO₃)₂ in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Strontium Standard, 1000 mg/L, VWR® AVS TITRINORM, Matrix: Nitric acid dil., Application: AAS standards
Supplier: VWR Chemicals
Description: Sr(NO₃)₂ in HNO₃ 2 to 5%.
Catalog Number: (84994.180)
Supplier: VWR Chemicals
Description: Strontium Standard, 1000 mg/L, VWR®, Matrix: H₂O, Application: Ion chromatography standards
UOM: 1 * 100 mL
Supplier: Thermo Scientific
Description: Strontium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: Thermo Scientific
Description: Strontium Standard, 10000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Catalog Number: (14205.18)
Supplier: Thermo Scientific
Description: Strontium Standard, Standard for wear metal analysis, 1000 µg/g, Specpure®, Matrix: oil, Application: Oil based standards
UOM: 1 * 50 g
Catalog Number: (PERCN9303802)
Supplier: PerkinElmer
Description: Strontium Standard, 1000 mg/L, Matrix: 2% HNO₃, Application: AAS standards, ICP standards
UOM: 1 * 1 items
Supplier: ACSD
Description: Strontium Standard, 1000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
Catalog Number: (ACSDM356.2NP.L1)
Supplier: ACSD
Description: Strontium Standard, 100 mg/L, Matrix: 2% HNO₃, Application: ICP-MS standards
UOM: 1 * 100 mL
Supplier: ACSD
Description: Strontium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: ACSD
Description: Strontium Standard, 1000 mg/L, Matrix: 2% HCl, Application: ICP standards
Supplier: Thermo Scientific
Description: Strontium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: AAS standards
Supplier: ACSD
Description: Strontium Standard, 10000 mg/L, Matrix: 2% HCl, Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 22
no targeter for Bottom