You Searched For: Silicium Standards
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Silicium Standard, 1000 mg/L, VWR® AVS TITRINORM, Silicon, Matrix: H₂O, Application: AAS standards
Supplier: VWR Chemicals
Description: (NH₄)₂ SiF₆ in H₂O tr. HF.
Supplier: VWR Chemicals
Description: Silicium Standard, 1000 mg/L, VWR® ARISTAR®, Silicon, Matrix: H₂O/tr HF, Application: ICP-MS standards
Catalog Number: (456012Y)
Supplier: VWR Chemicals
Description: (NH₄)₂ SiF₆ in H₂O tr. HF.
UOM: 1 * 100 mL
Catalog Number: (14209.18)
Supplier: Thermo Scientific
Description: Silicium Standard, Standard for wear metal analysis, 1000 µg/g, Specpure®, Matrix: oil, Application: Oil based standards
UOM: 1 * 50 g
Supplier: Thermo Scientific
Description: Silicium Standard, 10000 mg/L, Specpure®, Matrix: H₂O/tr HF, Application: ICP standards
Catalog Number: (43867.18)
Supplier: Thermo Scientific
Description: Silicium Standard, Standard for wear metal analysis, 5000 µg/g, Specpure®, Matrix: Mineral oil, Application: Oil based standards
UOM: 1 * 50 g
Supplier: Thermo Scientific
Description: Silicium Standard, 1000 mg/L, Specpure®, Matrix: H₂O/tr HF, Application: ICP standards
Catalog Number: (PERCN9303799)
Supplier: PerkinElmer
Description: Silicium Standard, 1000 mg/L, Matrix: Hâ‚‚O, Application: AAS standards, ICP standards
UOM: 1 * 125 mL
MSDS
Supplier: ACSD
Description: Silicium Standard, 10000 mg/L, Matrix: Hâ‚‚O, Application: ICP standards
Supplier: ACSD
Description: Silicium Standard, 1000 mg/L, Matrix: Hâ‚‚O, Application: ICP standards
Supplier: ACSD
Description: Silicium Standard, 10 mg/L, Matrix: Hâ‚‚O, Application: ICP-MS standards
Supplier: ACCUSTANDARD
Description: Silicium Standard, 1000 mg/L, Matrix: Hâ‚‚O/tr HF, Application: ICP standards
Supplier: ACCUSTANDARD
Description: Silicium Standard, 1000 mg/L, Matrix: Hâ‚‚O/tr HF, Application: AAS standards
Catalog Number: (PERCN9300150)
Supplier: PerkinElmer
Description: Silicium Standard, 1000 mg/L, Matrix: Hâ‚‚O, Application: ICP standards, AAS standards
UOM: 1 * 1 items
MSDS
Supplier: LGC Standards PROMOCHEM
Description: This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom