You Searched For: Multi-Element Standards
117  results were found
SearchResultCount:"117"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (85026.180)
Supplier: VWR Chemicals
Description: Multi-Element Standard, Multi-element calibration standard in dil. nitric acid for ICP-MS, VWR® ARISTAR®, Application: ICP-MS standards
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Multi-Element Standard, Alternate Metals II (4 components), VWR®, Ca 500 mg/l, Na 500 mg/l, K 100 mg/l, Mg 100 mg/l, Matrix: 2% HNO₃, Application: ICP standards
Catalog Number: (85006.186)
Supplier: VWR Chemicals
Description: 28 elements 100 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.
UOM: 1 * 100 mL
Catalog Number: (89186.180)
Supplier: VWR Chemicals
Description: Multi-Element Standard, Multi-element quality control standard 100 mg/L {Ag; Al; Ba; Bi; Ca; Cd; Co; Cr; Cu; Fe; Ga; Ge; In; K; Li; Mg; Mn; Mo; Na; Nb; Ni; P; Pb; Re; Sb; Si; Sn; Ta; Ti; V; W; Zn} in nitric acid 5% ARISTAR® for ICP, VWR® ARISTAR®, Application: ICP standards
UOM: 1 * 100 mL
Catalog Number: (85036.180)
Supplier: VWR Chemicals
Description: ICP solution of eight elements in 7% nitric acid.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Multi-Element Standard, ICP multi-element standard solution X, 23 elements, As 50 µg/L; B 100 µg/L; Ba 50 µg/L; Be 20 µg/L; Bi 10 µg/L; Ca 35000 µg/L; Cd 20 µg/L; Co 25 µg/L; Cr 20 µg/L; Cu 20 µg/L; Fe 100 µg/L; K 3000 µg/L; Mg 15000 µg/L; Mn 30 µg/L; Mo 100 µg/L; Na 8000 µg/L; Ni 50 µg/L; Pb 25 µg/L; Se 10 µg/L; Sr 100 µg/L; Tl 10 µg/L; V 50 µg/L; Zn 50 µg/L, VWR® ARISTAR®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: VWR Chemicals
Description: 6 components: Lithium (Li+) 50 mg/l, Sodium (Na+) 200 mg/l, Ammonium (NH4+) 250 mg/l, Potassium (K+) 500 mg/l, Calcium (Ca2+) 500 mg/l, Magnesium (Mg2...
Catalog Number: (85038.180)
Supplier: VWR Chemicals
Description: Six elements: 100 mg/L each of Ir; Pt; Os; Rh; Pd; Ru in HCl 15%.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: In 5% nitric acid solution.
Catalog Number: (05200.185)
Supplier: VWR Chemicals
Description: 28 elements 1 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.
UOM: 1 * 100 mL
Catalog Number: (85025.180)
Supplier: VWR Chemicals
Description: Multi-Element Standard, Multi-element calibration standard IV in dil. nitric acid for ICP, VWR® ARISTAR®, Application: ICP standards
UOM: 1 * 100 mL
Catalog Number: (84792.180)
Supplier: VWR Chemicals
Description: Multi-Element Standard, Multi-element quality control standard 100 mg/L {Au; Ir; Pd; Pt; Rh; Ru; Os; Sn; Te} ARISTAR® for ICP, VWR® ARISTAR®, Matrix: 10% HCl, Application: ICP standards
UOM: 1 * 100 mL
Catalog Number: (456602B)
Supplier: VWR Chemicals
Description: 25 Elements in 5% nitric acid solution.
UOM: 1 * 100 mL
Catalog Number: (458352K)
Supplier: VWR Chemicals
Description: Cation in dilute HNO₃.
UOM: 1 * 100 mL
Catalog Number: (456462H)
Supplier: VWR Chemicals
Description: In 2% nitric acid solution.
UOM: 1 * 100 mL
Catalog Number: (91181.260)
Supplier: VWR Chemicals
Description: Multi-Element Standard, NexION Setup Solution (8 Elements), VWR®, 1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U, Matrix: 1% HNO₃, Application: ICP-MS standards
UOM: 1 * 500 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 117
no targeter for Bottom