You Searched For: Lutetium Standards
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Lu₂O₃ in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Lutetium Standard, 10 mg/L, VWR® ARISTAR®, Lutetium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Lu₂O₃ in HNO₃ 2 to 5%.
Supplier: Thermo Scientific
Description: Lutetium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: AAS standards
Supplier: ACSD
Description: Lutetium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: ACSD
Description: Lutetium Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Supplier: Thermo Scientific
Description: Lutetium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: ACCUSTANDARD
Description: Lutetium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: AAS standards
Supplier: ACCUSTANDARD
Description: Lutetium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Catalog Number: (PERCN9300130)
Supplier: PerkinElmer
Description: Lutetium Standard, 1000 mg/L, Matrix: 2% HNO₃, Application: ICP standards, AAS standards
UOM: 1 * 1 items
Supplier: LGC Standards PROMOCHEM
Description: These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectro...
Supplier: ACCUSTANDARD
Description: Lutetium Standard, 10000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom