You Searched For: Gadolinium Standards
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Gadolinium Standard, 1000 mg/L, VWR® ARISTAR®, Gadolinium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Gadolinium Standard, 1000 mg/L, VWR® AVS TITRINORM, Matrix: 2% HNO₃, Application: AAS standards
Supplier: VWR Chemicals
Description: Gd₂O₃ in HNO₃ (2 to 5%).
Supplier: VWR Chemicals
Description: Gd₂O₃ in HNO₃ 2 to 5%.
Supplier: Thermo Scientific
Description: Gd₂O₃ in 5% HNO₃
Supplier: ACSD
Description: Gadolinium Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Supplier: ACSD
Description: Gadolinium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: Thermo Scientific
Description: Gadolinium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: AAS standards
Supplier: ACCUSTANDARD
Description: Gadolinium Standard, 10000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: LGC Standards PROMOCHEM
Description: The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectrosc...
Supplier: ACCUSTANDARD
Description: Gadolinium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: AAS standards
Supplier: ACCUSTANDARD
Description: Gadolinium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Catalog Number: (PERCN9300118)
Supplier: PerkinElmer
Description: Gadolinium Standard, 1000 mg/L, Matrix: 2% HNO₃, Application: ICP standards, AAS standards
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom