You Searched For: Chromium Standards
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Chromium Standard, 10000 mg/L, VWR® ARISTAR®, Chromium, Matrix: 2-5% HNO₃, Application: ICP standards
Supplier: VWR Chemicals
Description: Chromium Standard, 10 mg/L, VWR® ARISTAR®, Chromium, Matrix: 2% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Chromium Standard, 1000 mg/L, VWR® AVS TITRINORM, Chromium, Matrix: 2% HNO₃, Application: AAS standards
Catalog Number: (5727.0100)
Supplier: Avantor
Description: Chromium Standard, 10000 mg/L, J.T.Baker® BAKER INSTRA-ANALYZED®, J.T. Baker®, Matrix: 2% HNO₃, Application: ICP standards
UOM: 1 * 100 mL
Supplier: Thermo Scientific
Description: Chromium Standard, 10000 mg/L, Specpure®, Matrix: 5% HCl, Application: ICP standards
Supplier: Thermo Scientific
Description: Chromium Standard, 10000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: Thermo Scientific
Description: Chromium Standard, 1000 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
Supplier: Thermo Scientific
Description: Chromium Standard, 1000 µg/g, Specpure®, Matrix: H₂O, Application: Ion chromatography standards
Catalog Number: (42438.18)
Supplier: Thermo Scientific
Description: Chromium Standard, 5000 µg/g, Specpure®, Matrix: Mineral oil, Application: Oil based standards
UOM: 1 * 50 g
Catalog Number: (14222.18)
Supplier: Thermo Scientific
Description: Chromium Standard, 1000 µg/g, Specpure®, Matrix: Mineral oil, Application: Oil based standards
UOM: 1 * 50 g
Supplier: Hach
Description: Standard solution for accuracy checks in total chromium determinations.
Supplier: ACSD
Description: Chromium Standard, 1000 mg/L, Matrix: Nitric acid dil., Application: ICP standards
Supplier: Thermo Scientific
Description: Chromium Standard, 1000 mg/L, Specpure®, Matrix: 5% HCl, Application: ICP standards
Catalog Number: (45251.AE)
Supplier: Thermo Scientific
Description: Chromium Standard, 10 mg/L, Specpure®, Matrix: 5% HNO₃, Application: ICP standards
UOM: 1 * 100 mL
Supplier: ACSD
Description: Chromium Standard, 1000 mg/L, Matrix: 2% HCl, Application: ICP standards
Supplier: ACSD
Description: Chromium Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom