You Searched For: Ammonium Standards
14  results were found
SearchResultCount:"14"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, VWR®, Matrix: H₂O, Application: Chromatography standards, Ion chromatography standards
Supplier: VWR Chemicals
Description: Ammonium Standard, Ammonium (in N), 1000 mg/L, VWR®, Contents: NH₄Cl in H₂O, Application: Ion chromatography standards
Catalog Number: (88084.180)
Supplier: VWR Chemicals
Description: Ammonium Standard, Ammonium standard solution, 250 mg/L, VWR®, Matrix: H₂O
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, VWR®, Contents: NH₄Cl in H₂O, Application: Ion chromatography standards
Catalog Number: (35560.AE)
Supplier: Thermo Scientific
Description: Standard from (NHâ‚„)â‚‚SOâ‚„ in Hâ‚‚O
UOM: 1 * 100 mL
Supplier: ACSD
Description: Ammonium Standard, Ammonium-Nitrogen standard solution, 1000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
Supplier: ACSD
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
Catalog Number: (662-0383)
Supplier: WTW
Description: Ammonium Standard, Ammonium standard solution, 10000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
UOM: 1 * 1 L
Supplier: Thermo Orion
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, Orionâ„¢, Application: ISE Calibration
Supplier: LGC Standards PROMOCHEM
Description: These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aq...
Supplier: LGC Standards PROMOCHEM
Description: These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aq...
Catalog Number: (ACCUICNH4N1X1)
Supplier: ACCUSTANDARD
Description: Ammonium Standard, Ammonium-Nitrogen standard solution, 100 mg/L, Matrix: Hâ‚‚O, Application: Environmental standards
UOM: 1 * 100 mL
Catalog Number: (713-4073)
Supplier: WTW
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
UOM: 1 * 500 mL
Catalog Number: (ACCUICNH410X1)
Supplier: ACCUSTANDARD
Description: Ammonium Standard, Ammonium standard solution, 1000 mg/L, Matrix: Hâ‚‚O, Application: Ion chromatography standards
UOM: 1 * 100 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 14 of 14
no targeter for Bottom