You Searched For: Aluminium Standards
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Aluminium Standard, 1000 mg/L, VWR®, Aluminium, Matrix: 2% HCl, Application: AAS standards
Supplier: VWR Chemicals
Description: Al(NO₃)₃.9H₂O in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Aluminium Standard, 1000 mg/L, VWR® ARISTAR®, Aluminium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: Thermo Scientific
Description: Aluminium Standard, 10000 mg/L, Specpure®, Matrix: 5% HCl, Application: ICP standards, ICP-MS standards
Supplier: Thermo Scientific
Description: Aluminium Standard, 5000 µg/g, Specpure®, Aluminium, Matrix: Mineral oil, Application: Oil based standards
Supplier: Thermo Scientific
Description: Aluminium Standard, 1000 mg/L, Specpure®, Matrix: 5% HCl, Application: AAS standards
Catalog Number: (ACRO195805000)
Supplier: Thermo Scientific
Description: Aluminium Standard, 1000 mg/L, Matrix: 0,5 M HNO₃, Application: AAS standards
UOM: 1 * 500 mL
MSDS
Supplier: Hach
Description: Standard solution for accuracy checks in aluminium determinations.
Catalog Number: (PERCN9303726)
Supplier: PerkinElmer
Description: Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
UOM: 1 * 1 items
Supplier: ACSD
Description: Aluminium Standard, 1000 mg/L, Matrix: 2% HNO₃, Application: ICP standards
Supplier: ACSD
Description: Aluminium Standard, 1000 mg/L, Matrix: 2% HCl, Application: ICP standards
Catalog Number: (ACSDM302.2NP.L1)
Supplier: ACSD
Description: Aluminium Standard, 100 mg/L, Matrix: 2% HNO₃, Application: ICP-MS standards
UOM: 1 * 100 mL
MSDS
Supplier: ACSD
Description: Aluminium Standard, 10000 mg/L, Matrix: 5% HCl, Application: ICP standards
Supplier: ACSD
Description: Aluminium Standard, 10000 mg/L, Matrix: 5% HNO₃, Application: ICP standards
Supplier: LGC Standards PROMOCHEM
Description: The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectrosc...
Supplier: LGC Standards PROMOCHEM
Description: The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectrosc...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom