You Searched For: Succinate Standards
2  results were found
SearchResultCount:"2"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (84995.180)
Supplier: VWR Chemicals
Description: Succinate standard solution, 1000 mg/l, VWR®, Matrix: H₂O, Application: Ion chromatography standards
UOM: 1 * 100 mL
Supplier: ACSD
Description: Succinate standard solution, 1000 mg/l, Matrix: Hâ‚‚O
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 2 of 2
no targeter for Bottom