You Searched For: Sulphate Standards
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (88090.180)
Supplier: VWR Chemicals
Description: Standard solution (1000 ppm SO₄), 1000 mg/l, VWR®, Contents: K₂SO₄ in H₂O, Application: Pharmaceutical standards
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Sulphate standard solution, 1000 mg/l, VWR®, Contents: Na₂SO₄ in H₂O, Application: Ion chromatography standards
Catalog Number: (88091.180)
Supplier: VWR Chemicals
Description: Standard solution (1000 ppm SO₄) R1 Reag. Ph. Eur. 5002801, 1000 mg/l, VWR®, Contents: K₂SO₄ in H₂O, Application: Pharmaceutical standards
UOM: 1 * 100 mL
Supplier: Thermo Scientific
Description: Sulphate standard solution, Specpure®, 1000 mg/l
Supplier: ACSD
Description: Sulphate standard solution, 1000 mg/l, Matrix: H₂O
Catalog Number: (662-0848)
Supplier: Thermo Orion
Description: Sulphate standard for ISE calibration, Orion™, 0,1M
UOM: 1 * 475 mL
Supplier: LGC Standards PROMOCHEM
Description: This standard solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and technique...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom