You Searched For: Cationic Standards
1 474  results were found
SearchResultCount:"1474"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (91181.260)
Supplier: VWR Chemicals
Description: Multi-Element Standard, NexION Setup Solution (8 Elements), VWR®, 1 ug/l each of Be, Ce, Fe, in, Li, Mg, Pb, U, Matrix: 1% HNO₃, Application: ICP-MS standards
UOM: 1 * 500 mL
Supplier: VWR Chemicals
Description: NHâ‚„Hâ‚‚POâ‚„ in Hâ‚‚O.
Catalog Number: (456462H)
Supplier: VWR Chemicals
Description: In 2% nitric acid solution.
UOM: 1 * 100 mL
Catalog Number: (458352K)
Supplier: VWR Chemicals
Description: Cation in dilute HNO₃.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Ytterbium Standard, 1000 mg/L, VWR® ARISTAR®, Ytterbium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Beryllium Standard, 1000 mg/L, VWR® ARISTAR®, Beryllium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Zinc Standard, 1000 mg/L, VWR®, Zinc, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: Gadolinium Standard, 1000 mg/L, VWR® AVS TITRINORM, Matrix: 2% HNO₃, Application: AAS standards
Supplier: VWR Chemicals
Description: Zn in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Potassium Standard, 10 ppm, VWR®, Potassium, Matrix: 2-5% HNO₃, Application: ICP-MS standards
Supplier: VWR Chemicals
Description: (NH₄)₂ TiF₆ in H₂O tr. HF.
Catalog Number: (6925.0100)
Supplier: Avantor
Description: Calcium Standard, 1000 mg/L, J.T.Baker®, J.T. Baker®, Matrix: Nitric acid dil., Application: AAS standards
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: KNO₃ in HNO₃ 2 to 5%.
Supplier: VWR Chemicals
Description: Strontium Standard, 1000 mg/L, VWR® AVS TITRINORM, Strontium, Matrix: Nitric acid dil., Application: AAS standards
Supplier: VWR Chemicals
Description: Tantalum Standard, 10000 mg/L, VWR® ARISTAR®, Tantalum, Matrix: H₂O/tr HF, Application: ICP standards
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 1 474
no targeter for Bottom