Sterile polycarbonate flasks with weldable tubing provide the maximum amount of flexibility and reliability.
- Sterile flasks come individually bagged and process ready
- Multiple flask sizes allow for process-specific flexibility
- When used with sterile VWR® tube sets, custom possibilities are endless
- Gamma irradiated to Sterility Assurance Level (SAL) 10-6
These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They are offered in a comprehensive size selection with tubing that is completely weldable/sealable. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to 65 °C without losing integrity. All assemblies are individually double-bagged with product label located on inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6. Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications.