You Searched For: Standard Laboratory Blenders
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: WARING BLENDOR
Description: Reliable devices with powerful motors. For thorough mixing, dispersion and emulsification. Special cutter heads and high speed operation make these la...
Supplier: WARING BLENDOR
Description: The CB15 series blenders are designed specifically for processing large quantities of free flowing material with a high liquid content. This range of ...
Catalog Number: (GERB7610)
Supplier: GERBER FUNKE DR N.
Description: Solubility index mixer is used for determining the solubility of milk, cream and whey powder.
UOM: 1 * 1 items
Catalog Number: (WARICAC139)
Supplier: WARING BLENDOR
Description: The CAC139 is the clear copolyester 1,36 L blender container from Waring with the blade assembly included. These containers are designed for the TORQ ...
UOM: 1 * 1 items
New Product
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom