Classic socks, SK13
Supplier: Portwest
Certificates
About this item
Informal business socks made from 80% fine combed cotton and 20% nylon.
- With cushioned sole area for comfort and durability
Height: 25 cm
Weight: 60 g
Specifications
- Catalog No:
- 114-7159
- 114-7160
- Model:
- SK13
- SK13
- Garment application:
- Industrial
- Industrial
- Gender:
- Unisex
- Unisex
- Fabric:
- 80% cotton and 20% nylon
- 80% cotton and 20% nylon
- Weight:
- 60 g
- 60 g
Specifications
Product Information
- Catalog #Supplier #SeqTypeColourGarment fitShoe sizePkAvailabilityPrice
- 114-7159SK13BKR39-4301Classic socksBlackRegular39 - 43 (6 - 9)1 PAIRLog in for availability1 * 1 PAIR5,00 €Specifications:
More
Specifications
ModelSK13Garment applicationIndustrialEnvironmentally PreferableRisk areaGenderUnisexFabric80% cotton and 20% nylonWeight60 g - 114-7160SK13BKR44-4802Classic socksBlackRegular44 - 48 (10 - 13)1 PAIRLog in for availability1 * 1 PAIR5,00 €Specifications:
More
Specifications
ModelSK13Garment applicationIndustrialEnvironmentally PreferableRisk areaGenderUnisexFabric80% cotton and 20% nylonWeight60 g