This exercise tubing is produced with a multi-dipping latex manufacturing technology to attain the highest quality possible. The tubing provides an exceptionally good breakage resistance. Ideal for use in physiotherapy and rehabilitation as well as for fitness and sports. The tubes should ideally be used with handles (available separately).
- Easy to use
- Long-lasting and durable
- Eco-friendly: Made of biodegradable natural latex
- Decrease of the natural adhesive latex protein: Reduction of the allergenic potential
- 35 m roll enables users to choose the exact right length for their specific application
- Available in different resistance levels (colour-coded)
Certifikace: TÜV-GS certified
Informace pro objednávání: Supplied without handles which can be ordered separately.