Capillary columns, PEG-20M
: Agilent
Stainless steel
Product Information
- Catalog #Supplier #SeqColumn FormatØ int.LengthMesh sizePkAvailabilityPrice
- 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-80122G3591-8012202Capillary2,00 mm2 m80/10011 * 1 itemsSpecifications:
Specifications
Environmentally Preferable - 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-80123G3591-8012303Capillary2,00 mm4 m80/10011 * 1 items470,00 €
Specifications
Environmentally Preferable - 10% PEG-20M, Chromosorb W support10% PEG-20M, Chromosorb W supportHEWLG3591-80119G3591-8011901Capillary2,00 mm2 m80/10011 * 1 items333,00 €
Specifications
Environmentally Preferable
UltiMetal
- SeqColumn FormatØ int.LengthMesh sizePk
- 10% PEG-20M, Chromosorb W support10% PEG-20M, Chromosorb W supportHEWLG3591-81119G3591-8111904Capillary2,00 mm2 m80/10011 * 1 items333,00 €
Specifications
Environmentally Preferable - 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-81122G3591-8112205Capillary2,00 mm2 m80/10011 * 1 items333,00 €
Specifications
Environmentally Preferable - 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-81123G3591-8112306Capillary2,00 mm4 m80/10011 * 1 items
Specifications
Environmentally Preferable
Nickel
- SeqColumn FormatØ int.LengthMesh sizePk
- 10% PEG-20M, Chromosorb W support10% PEG-20M, Chromosorb W supportHEWLG3591-82119G3591-8211907Capillary2,00 mm2 m80/10011 * 1 items407,00 €
Specifications
Environmentally Preferable - 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-82122G3591-8212208Capillary2,00 mm2 m80/10011 * 1 items
Specifications
Environmentally Preferable - 20% PEG-20M, Chromosorb W support20% PEG-20M, Chromosorb W supportHEWLG3591-82123G3591-8212309Capillary2,00 mm4 m80/10011 * 1 items