Digital Solutions
Applications
Support
About
$0.00
Order Entry
MES-SDS running buffer (20X)
:
Thermo Fisher Scientific
MES-SDS running buffer (20X)
:
Thermo Fisher Scientific
Physical Form: Liquid
Environmentally Preferable:
msds_ALFAJ62138
Thermo Scientific Acroseal 25 Years
Pk
MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
J62138.AP
J62138.AP
500 mL
1 * 500 mL
1 * 500 mL - €118.00
€118.00
-
+
1 * 500 mL
€118.00
-
+
MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
J62138.K2
J62138.K2
1 L
1 * 1 L
1 * 1 L - €223.00
€223.00
-
+
1 * 1 L
€223.00
-
+