VWR® HazMat, Absorbent mat pads and rolls
Supplier: Avantor
VWR®
VWRU82028-024EA 0 EUR
VWRU89169-000 VWRU89169-002 VWRU82028-024 VWRU82028-026
VWR® HazMat, Absorbent mat pads and rolls
Spill Control Materials
Polypropylene absorbent pads and rolls for spill cleanup; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety.
  • Soak up leaks and spills fast, keeping floors at your workplace clean, dry, and safe
  • High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
  • Can be incinerated after use to reduce waste
  • Efficient high-absorbency materials reduce waste of absorbents: Hazmat pads and rolls are designed for aggressive fluids such as acids, caustics, and bases.
Efficient high-absorbency materials reduce waste of absorbents: Hazmat pads and rolls are designed for aggressive fluids such as acids, caustics, and bases.