Autoradiography cassettes, BioMax®
Supplier: Carestream Health
730-0420EA 784 EUR
730-0420
Autoradiography cassettes, BioMax®
Autoradiography Films Cassettes
These BioMax® cassettes are designed to increase image resolution and clarity of the background ensuring intimate contact between the film and the sample in an environment protected from light.
When using for low temperature exposures (–70 °C), the cassettes are to be allowed to warm to room temperature before opening to prevent the formation of condensation and damage related artifacts hinges of the cassette.