DNA ladder, 500 bp, DNAmark™
Supplier: G-Biosciences
GENO786-462EA 150 EUR
GENO786-462
DNA ladder, 500 bp, DNAmark™
Electrophoresis Reagents Molecular Weight Markers
DNAmark™ 500 bp ladder consists of 10 DNA fragments ranging from 500 bp to 5000 bp. The 2000 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 2000 bp are 40 ng. The 2000 bp fragment is 100 ng.
  • Pre-mixed with loading dye and ready to use