Plasmid Miniprep Kit I, peqGOLD
PEQL12-6942-00EA 16.3 EUR
PEQL12-6942-00 PEQLEMR500050 PEQL12-6942-01 PEQL12-6942-02
Plasmid Miniprep Kit I, peqGOLD
Plasmid Miniprep Kit I for the easy, quick and efficient extraction of highly pure plasmid DNA. Yields of up to 25 µg super coiled plasmid DNA per preparation. Plasmid DNA obtained with peqGOLD Plasmid Miniprep kits can directly be used for a wide range of downstream applications.
  • Suitable for the simultaneous processing of multiple samples
  • Short hands-on and preparation times (<15 minutes)
  • No need for organic extractions or precipitations
  • Complete enzymatic degradation of all cellular RNA
  • Efficient removal of enzyme inhibitors and contaminants
  • Final elution in Elution Buffer or water
*Safety Line columns with snap-on lids for maximum protection against cross-contamination. Classic Line columns without lids for easy handling.
Delivery information: Each kit includes PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, RNase A Solution, Solution I, II and III, PW Plasmid and DNA Wash Buffer and Elution Buffer.